siRNA Details
| Gene Symbol: NPSR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NR_015356.2 | 3 | 329 | |||
| NR_033664.1 | 2 | 258 | ||||
| NR_033665.1 | 2 | 258 | ||||
| GenBank mRNA | AY312364.1 | - | 3 | 332 | ||
| AY312365.1 | - | LIT3317 AAA1 |
3 | 327 | ||
| AY312366.1 | - | LIT3326 AAA1 |
3 | 223 | ||
| AY312367.1 | - | LIT3327 AAA1 |
2 | 258 | ||
| AY312368.1 | - | LIT3328 AAA1 |
2 | 258 | ||
| AY312369.1 | - | LIT3332 AAA1 |
2 | 258 | ||
| AY312370.1 | - | LIT3333 AAA1 |
2 | 143 | ||
| AY312371.1 | - | LIT3334 AAA1 |
2 | 258 | ||
| AY312372.1 | - | LIT3335 AAA1 |
2 | 258 | ||
| AY312373.1 | - | LIT3331 AAA1 |
3 | 223 | ||
| AY321515.1 | - | 2 | 139 | |||
| AY321516.1 | - | 2 | 139 | |||
| BC031961.1 | - | 3 | 310 | |||
| BC130477.1 | - | 2 | 143 | |||
| BX649082.1 | - | 2 | 221 | |||
| U85653.1 | - | 2 | 221 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG723002.1 | - | 3 | 315 | Chr. 7 - 34719597 - 34834332 [-] on Build GRCh38 | |

NPSR1-AS1
Genomic Map