siRNA Details
| Gene Symbol: LOC100129940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292023.1 | NP_001278952.1 | 3 | 1834 | ||
| GenBank mRNA | AK128707.1 | - | 3 | 1708 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100129940| Gene Symbol: LOC100129940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001292023.1 | NP_001278952.1 | 3 | 1834 | ||
| GenBank mRNA | AK128707.1 | - | 3 | 1708 | ||