siRNA Details
| Gene Symbol: LOC102724190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110553.1 | 3 | 307 | |||
| GenBank mRNA | AK124327.1 | - | 3 | 307 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB245360.1 | - | 3 | 307 | Chr. 14 - 77070427 - 77076191 [-] on Build GRCh38 | |

LOC102724190
Genomic Map