siRNA Details
| Gene Symbol: LOC102724190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110553.1 | 3 | 1592 | |||
| GenBank mRNA | AK124327.1 | - | 3 | 1592 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB326058.1 | - | 1 | 99 | Chr. 14 - 77069001 - 77069503 [-] on Build GRCh38 | |

LOC102724190
Genomic Map