siRNA Details
Gene Symbol: LINC01492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121578.1 | 10 | 2317 | |||
GenBank mRNA | BC035187.1 | - | 10 | 2324 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121578.1 | 10 | 2317 | |||
GenBank mRNA | BC035187.1 | - | 10 | 2324 |