siRNA Details
Gene Symbol: TAPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027696.1 | 3 | 1064 | |||
NR_027697.1 | 3 | 1013 | ||||
GenBank mRNA | DA797317.1 | - | 1 | 329 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA778084.1 | - | 3 | 605 | Chr. 4 - 16227071 - 16256744 [+] on Build GRCh38 |
