siRNA Details
| Gene Symbol: LOC101929486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109868.1 | 12 | 2843 | |||
| GenBank mRNA | BC036700.1 | - | 12 | 2847 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929486| Gene Symbol: LOC101929486 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109868.1 | 12 | 2843 | |||
| GenBank mRNA | BC036700.1 | - | 12 | 2847 | ||