siRNA Details
| Gene Symbol: LOC101929116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121584.1 | 3 | 1159 | |||
| GenBank mRNA | BC031239.1 | - | 3 | 1161 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929116| Gene Symbol: LOC101929116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121584.1 | 3 | 1159 | |||
| GenBank mRNA | BC031239.1 | - | 3 | 1161 | ||