siRNA Details
| Gene Symbol: CASC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015410.1 | 7 | 1036 | |||
| GenBank mRNA | AK022865.1 | - | 6 | 662 | ||
| AK026189.1 | - | 7 | 1036 | |||
| BC044235.1 | - | 4 | 485 | |||
| KP981381.1 | - | 4 | 550 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM564069.1 | - | 499 | Chr. - - [-] on Build | ||

CASC15
Genomic Map