siRNA Details
| Gene Symbol: CASC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015410.1 | 11 | 1498 | |||
| GenBank mRNA | AK022865.1 | - | 11 | 1245 | ||
| AK026189.1 | - | 11 | 1498 | |||
| AK094718.1 | - | 3 | 746 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASC15| Gene Symbol: CASC15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015410.1 | 11 | 1498 | |||
| GenBank mRNA | AK022865.1 | - | 11 | 1245 | ||
| AK026189.1 | - | 11 | 1498 | |||
| AK094718.1 | - | 3 | 746 | |||