siRNA Details
| Gene Symbol: LOC439933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122079.1 | 5 | 597 | |||
| NR_122080.1 | 3 | 472 | ||||
| GenBank mRNA | BC030111.1 | - | 3 | 478 | ||
| BC037904.2 | - | 5 | 597 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC439933| Gene Symbol: LOC439933 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122079.1 | 5 | 597 | |||
| NR_122080.1 | 3 | 472 | ||||
| GenBank mRNA | BC030111.1 | - | 3 | 478 | ||
| BC037904.2 | - | 5 | 597 | |||