siRNA Details
| Gene Symbol: LINC01510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120506.1 | 5 | 934 | |||
| GenBank mRNA | BG189453.1 | - | 2 | 269 | ||
| BG195638.1 | - | 2 | 268 | |||
| BG213277.1 | - | 2 | 271 | |||
| LN812953.1 | - | 4 | 595 | |||
| LN812954.1 | - | 5 | 899 | |||

LINC01510
Genomic Map