siRNA Details
| Gene Symbol: ZFPM2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125796.1 | 5 | 462 | |||
| GenBank mRNA | BF977562.1 | - | 349 | |||
| BU959299.1 | - | 437 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZFPM2-AS1| Gene Symbol: ZFPM2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125796.1 | 5 | 462 | |||
| GenBank mRNA | BF977562.1 | - | 349 | |||
| BU959299.1 | - | 437 | ||||