siRNA Details
Gene Symbol: LOC101927165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135590.1 | 2 | 88 | |||
GenBank mRNA | BC040311.1 | - | 2 | 85 | ||
DB084887.1 | - | 2 | 88 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135590.1 | 2 | 88 | |||
GenBank mRNA | BC040311.1 | - | 2 | 85 | ||
DB084887.1 | - | 2 | 88 |