siRNA Details
| Gene Symbol: SDCBP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040047.1 | 3 | 1345 | |||
| NR_040048.1 | 4 | 1307 | ||||
| NR_040049.1 | 3 | 1092 | ||||
| GenBank mRNA | AK055993.1 | - | 4 | 1229 | ||
| BC034773.1 | - | 3 | 1025 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SDCBP2-AS1| Gene Symbol: SDCBP2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040047.1 | 3 | 1345 | |||
| NR_040048.1 | 4 | 1307 | ||||
| NR_040049.1 | 3 | 1092 | ||||
| GenBank mRNA | AK055993.1 | - | 4 | 1229 | ||
| BC034773.1 | - | 3 | 1025 | |||