siRNA Details
Gene Symbol: LOC101930452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120479.1 | 1 | 1161 | |||
GenBank mRNA | BC040314.1 | - | 1 | 1137 | ||
BC047234.1 | - | 1 | 1161 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101930452 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120479.1 | 1 | 1161 | |||
GenBank mRNA | BC040314.1 | - | 1 | 1137 | ||
BC047234.1 | - | 1 | 1161 |