siRNA Details
| Gene Symbol: SLC39A12-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038419.1 | 5 | 784 | |||
| GenBank mRNA | AK055012.1 | - | 5 | 784 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC39A12-AS1| Gene Symbol: SLC39A12-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038419.1 | 5 | 784 | |||
| GenBank mRNA | AK055012.1 | - | 5 | 784 | ||