siRNA Details
| Gene Symbol: LINC00494 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026958.1 | 5 | 2214 | |||
| GenBank mRNA | AL832259.1 | - | 3 | 1702 | ||
| BC035164.2 | - | 5 | 2178 | |||
| BC122555.1 | - | 1 | 583 | 40084813 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK129540.1 | - | 1 | 568 | Chr. 20 - 47976762 - 47978122 [-] on Build GRCh38 | |

LINC00494
Genomic Map