siRNA Details
| Gene Symbol: IGSF11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046230.1 | 1, 2 | 114 | |||
| GenBank mRNA | BI829854.1 | - | 1, 2 | 114 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

IGSF11-AS1| Gene Symbol: IGSF11-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046230.1 | 1, 2 | 114 | |||
| GenBank mRNA | BI829854.1 | - | 1, 2 | 114 | ||