siRNA Details
| Gene Symbol: LOC101929697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134939.1 | 2 | 280 | |||
| NR_134940.1 | 2 | 244 | ||||
| GenBank mRNA | BI042007.1 | - | 2 | 298 | ||
| DA288418.1 | - | 2 | 247 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929697| Gene Symbol: LOC101929697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134939.1 | 2 | 280 | |||
| NR_134940.1 | 2 | 244 | ||||
| GenBank mRNA | BI042007.1 | - | 2 | 298 | ||
| DA288418.1 | - | 2 | 247 | |||