siRNA Details
| Gene Symbol: DUBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028302.1 | 1, 2 | 394 | |||
| GenBank mRNA | AK309100.1 | - | 1, 2 | 394 | ||
| DA339707.1 | - | 1, 2 | 394 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DUBR| Gene Symbol: DUBR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028302.1 | 1, 2 | 394 | |||
| GenBank mRNA | AK309100.1 | - | 1, 2 | 394 | ||
| DA339707.1 | - | 1, 2 | 394 | |||