siRNA Details
Gene Symbol: LINC00488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026767.1 | 3 | 915 | |||
GenBank mRNA | AK123226.1 | - | 3 | 915 | ||
BC022563.1 | - | 3 | 904 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026767.1 | 3 | 915 | |||
GenBank mRNA | AK123226.1 | - | 3 | 915 | ||
BC022563.1 | - | 3 | 904 |