siRNA Details
| Gene Symbol: LINC00488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026767.1 | 3 | 752 | |||
| GenBank mRNA | AK123226.1 | - | 3 | 752 | ||
| BC022563.1 | - | 3 | 742 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI763241.1 | - | 3 | 726 | Chr. 3 - 109178190 - 109183513 [+] on Build GRCh38 | |

LINC00488
Genomic Map