siRNA Details
Gene Symbol: LOC101927196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110214.1 | 4 | 583 | |||
NR_110215.1 | 4 | 541 | ||||
NR_110216.1 | 4 | 546 | ||||
NR_110217.1 | 3 | 419 | ||||
GenBank mRNA | BC039382.1 | - | 4 | 539 | ||
BU561233.1 | - | 3 | 422 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI461459.1 | - | 4 | 544 | Chr. 2 - 185720236 - 185738721 [-] on Build GRCh38 |
