siRNA Details
| Gene Symbol: LINC00606 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027102.1 | 2 | 305 | |||
| GenBank mRNA | AK093398.1 | - | 2 | 305 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB034700.1 | - | 2 | 305 | Chr. 3 - 10761767 - 10764192 [-] on Build GRCh38 | |

LINC00606
Genomic Map