siRNA Details
| Gene Symbol: LINC00880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034007.1 | 4 | 1359 | |||
| GenBank mRNA | BC040669.1 | - | 4 | 1354 | ||
| BC122546.1 | - | 1 | 959 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00880| Gene Symbol: LINC00880 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034007.1 | 4 | 1359 | |||
| GenBank mRNA | BC040669.1 | - | 4 | 1354 | ||
| BC122546.1 | - | 1 | 959 | |||