siRNA Details
Gene Symbol: LINC00880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034007.1 | 4 | 436 | |||
GenBank mRNA | AK311218.1 | - | 4 | 438 | ||
BC040669.1 | - | 4 | 432 | |||
BC122546.1 | - | 1 | 38 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034007.1 | 4 | 436 | |||
GenBank mRNA | AK311218.1 | - | 4 | 438 | ||
BC040669.1 | - | 4 | 432 | |||
BC122546.1 | - | 1 | 38 |