siRNA Details
Gene Symbol: LOC388882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110539.1 | 10 | 922 | |||
GenBank mRNA | BC036910.1 | - | 10 | 922 | 5164270 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN270671.1 | - | 6 | 659 | Chr. 22 - 23467770 - 23473950 [-] on Build GRCh38 |
