siRNA Details
Gene Symbol: LOC388882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110539.1 | 12 | 1553 | |||
GenBank mRNA | BC036910.1 | - | 12 | 1573 | 5164270 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC388882 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110539.1 | 12 | 1553 | |||
GenBank mRNA | BC036910.1 | - | 12 | 1573 | 5164270 |