siRNA Details
Gene Symbol: LOC105377348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131185.1 | 3 | 859 | |||
NR_131186.1 | 2 | 729 | ||||
GenBank mRNA | AK093356.1 | - | 2 | 650 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI830848.1 | - | 2 | 637 | Chr. 4 - 102828150 - 102844062 [+] on Build GRCh38 |
