siRNA Details
| Gene Symbol: LOC101926911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110104.1 | 1 | 70 | |||
| GenBank mRNA | AK055351.1 | - | 1 | 70 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DA438346.1 | - | 1 | 73 | Chr. 15 - 91022619 - 91030706 [+] on Build GRCh38 | |

LOC101926911
Genomic Map