siRNA Details
Gene Symbol: CCDC37-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103787.1 | 3 | 900 | |||
NR_103788.1 | 3 | 781 | ||||
NR_103789.1 | 2 | 729 | ||||
GenBank mRNA | BC033989.2 | - | 3 | 863 | ||
BX109262.1 | - | 3 | 526 |
