siRNA Details
| Gene Symbol: LOC105375843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134323.1 | 2 | 495 | |||
| GenBank mRNA | AA412543.1 | - | 2 | 495 | ||
| CR738863.1 | - | 2 | 494 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105375843| Gene Symbol: LOC105375843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134323.1 | 2 | 495 | |||
| GenBank mRNA | AA412543.1 | - | 2 | 495 | ||
| CR738863.1 | - | 2 | 494 | |||