siRNA Details
| Gene Symbol: LOC101927331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135589.1 | 2 | 109 | |||
| GenBank mRNA | BC014368.1 | - | 2 | 110 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927331| Gene Symbol: LOC101927331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135589.1 | 2 | 109 | |||
| GenBank mRNA | BC014368.1 | - | 2 | 110 | ||