siRNA Details
| Gene Symbol: LOC101928445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110560.1 | 2 | 376 | |||
| GenBank mRNA | AK128666.1 | - | 2 | 376 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB258449.1 | - | 2 | 376 | Chr. 5 - 179602247 - 179603741 [-] on Build GRCh38 | |

LOC101928445
Genomic Map