siRNA Details
| Gene Symbol: ZBED3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024398.1 | 4 | 806 | |||
| GenBank mRNA | AK055144.1 | - | 5 | 1229 | ||
| AK098256.1 | - | 3 | 1476 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBED3-AS1| Gene Symbol: ZBED3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024398.1 | 4 | 806 | |||
| GenBank mRNA | AK055144.1 | - | 5 | 1229 | ||
| AK098256.1 | - | 3 | 1476 | |||