siRNA Details
| Gene Symbol: LOC101928137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110130.1 | 4 | 769 | |||
| GenBank mRNA | AK310448.1 | - | 4 | 771 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928137| Gene Symbol: LOC101928137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110130.1 | 4 | 769 | |||
| GenBank mRNA | AK310448.1 | - | 4 | 771 | ||