siRNA Details
| Gene Symbol: LINC00637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038436.1 | 2 | 109 | |||
| GenBank mRNA | AK057986.1 | - | 2 | 111 | ||
| BC112204.1 | - | 1 | 83 | |||
| BC112206.1 | - | 1 | 83 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00637| Gene Symbol: LINC00637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038436.1 | 2 | 109 | |||
| GenBank mRNA | AK057986.1 | - | 2 | 111 | ||
| BC112204.1 | - | 1 | 83 | |||
| BC112206.1 | - | 1 | 83 | |||