siRNA Details
| Gene Symbol: LOC101928673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125960.1 | 2 | 1159 | |||
| GenBank mRNA | AK130460.1 | - | 2 | 1160 | ||
| BC136808.1 | - | 2 | 1152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101928673| Gene Symbol: LOC101928673 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125960.1 | 2 | 1159 | |||
| GenBank mRNA | AK130460.1 | - | 2 | 1160 | ||
| BC136808.1 | - | 2 | 1152 | |||