siRNA Details
| Gene Symbol: LOC100507534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110649.1 | 1 | 93 | |||
| GenBank mRNA | BC039433.1 | - | 1 | 94 | ||
| BC048130.1 | - | 1 | 94 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG708079.1 | - | 1 | 98 | Chr. 16 - 47849903 - 47887131 [-] on Build GRCh38 | |

LOC100507534
Genomic Map