siRNA Details
| Gene Symbol: LINC01049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120414.1 | 6 | 567 | |||
| GenBank mRNA | BC038529.1 | - | 6 | 567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01049| Gene Symbol: LINC01049 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120414.1 | 6 | 567 | |||
| GenBank mRNA | BC038529.1 | - | 6 | 567 | ||