siRNA Details
| Gene Symbol: LINC01393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120521.1 | 2 | 171 | |||
| GenBank mRNA | BC022431.1 | - | 2 | 171 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01393| Gene Symbol: LINC01393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120521.1 | 2 | 171 | |||
| GenBank mRNA | BC022431.1 | - | 2 | 171 | ||