siRNA Details
| Gene Symbol: LOC101927755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110815.1 | 4 | 497 | |||
| GenBank mRNA | AK097658.1 | - | 4 | 497 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB044175.1 | - | 4 | 497 | Chr. 17 - 59965096 - 59995926 [+] on Build GRCh38 | |

LOC101927755
Genomic Map