siRNA Details
| Gene Symbol: MIRLET7DHG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046163.1 | 4 | 214 | |||
| NR_046165.1 | 3 | 155 | ||||
| GenBank mRNA | BC036695.1 | - | 4 | 193 | ||
| BC045813.1 | - | 3 | 155 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIRLET7DHG| Gene Symbol: MIRLET7DHG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046163.1 | 4 | 214 | |||
| NR_046165.1 | 3 | 155 | ||||
| GenBank mRNA | BC036695.1 | - | 4 | 193 | ||
| BC045813.1 | - | 3 | 155 | |||