siRNA Details
| Gene Symbol: LINC01359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119383.1 | 5 | 742 | |||
| GenBank mRNA | BC035370.1 | - | 5 | 742 | ||
| BI825893.1 | - | 5 | 744 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01359| Gene Symbol: LINC01359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119383.1 | 5 | 742 | |||
| GenBank mRNA | BC035370.1 | - | 5 | 742 | ||
| BI825893.1 | - | 5 | 744 | |||