siRNA Details
| Gene Symbol: LINC01359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_119383.1 | 2 | 173 | |||
| GenBank mRNA | BC035370.1 | - | 2 | 173 | ||
| BI825893.1 | - | 2 | 173 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BI826506.1 | - | 2 | 173 | Chr. 1 - 64987461 - 65002475 [-] on Build GRCh38 | |

LINC01359
Genomic Map