siRNA Details
| Gene Symbol: LOC100507144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120528.1 | 1, 2 | 328 | |||
| GenBank mRNA | AW291715.1 | - | 1, 2 | 341 | ||
| BM764748.1 | - | 1, 2 | 178 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100507144| Gene Symbol: LOC100507144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120528.1 | 1, 2 | 328 | |||
| GenBank mRNA | AW291715.1 | - | 1, 2 | 341 | ||
| BM764748.1 | - | 1, 2 | 178 | |||