siRNA Details
| Gene Symbol: KDM4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098534.1 | - | 3 | 1194 | ||
| BC042976.2 | - | 3 | 1248 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KDM4C| Gene Symbol: KDM4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK098534.1 | - | 3 | 1194 | ||
| BC042976.2 | - | 3 | 1248 | |||