siRNA Details
| Gene Symbol: LIFR-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103553.1 | 1 | 1267 | |||
| NR_103554.1 | 1 | 1267 | ||||
| GenBank mRNA | BC038371.1 | - | 1 | 1267 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIFR-AS1| Gene Symbol: LIFR-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_103553.1 | 1 | 1267 | |||
| NR_103554.1 | 1 | 1267 | ||||
| GenBank mRNA | BC038371.1 | - | 1 | 1267 | ||