siRNA Details
| Gene Symbol: ESR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017021080.1 | XP_016876569.1 | 2 | 762 | ||
| XM_017021082.1 | XP_016876571.1 | 1 | 390 | |||
| GenBank mRNA | BX247957.1 | - | 2 | 730 | ||
| KC777381.1 | - | 1 | 29 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ESR2| Gene Symbol: ESR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017021080.1 | XP_016876569.1 | 2 | 762 | ||
| XM_017021082.1 | XP_016876571.1 | 1 | 390 | |||
| GenBank mRNA | BX247957.1 | - | 2 | 730 | ||
| KC777381.1 | - | 1 | 29 | |||